Novus Biologicals
Manufacturer Code:NBP177342
Catalog # NBP177342
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 15 amino acid synthetic peptide near the center of human TOM70. The immunogen is located within amino acids 220 - 270 of TOM70. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: FLJ90470 KIAA0719 mitochondrial import receptor subunit TOM70 Mitochondrial precursor proteins import receptor TOM70 Translocase of outer membrane 70 kDa subunit translocase of outer mitochondrial membrane 70 (yeast) homolog A translocase of outer mitochondrial membrane 70 homolog A (S. cerevisiae) translocase of outer mitochondrial membrane 70 homolog A (yeast); Mitochondrial import receptor subunit TOM70; Mitochondrial precursor proteins import receptor; Translocase of outer membrane 70 kDa subunit; translocase of outer mitochondrial membrane 70 homolog A; Translocase of outer mitochondrial membrane protein 70
Gene Aliases: KIAA0719; TOM70; TOMM70; TOMM70A
UniProt ID: (Human) O94826
Entrez Gene ID: (Human) 9868
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.