Proteintech
Manufacturer Code:149151AP150UL
Catalog # 50-172-9831
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TOB1 Fusion Protein Ag6714 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APRO6 PIG49 Protein Tob1 TOB TOB1 Transducer of erbB 2 1 transducer of ERBB2 1 TROB TROB1; proliferation-inducing gene 49; Protein Tob1; Transducer of erbB-2 1; transducer of erbB2; transducer of ERBB2, 1
Gene Aliases: APRO5; APRO6; PIG49; TOB; TOB1; TROB; TROB1
UniProt ID: (Human) P50616, (Mouse) Q61471
Entrez Gene ID: (Human) 10140, (Mouse) 22057, (Rat) 170842
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.