OriGene
Manufacturer Code:UM800097CF
Catalog # 50-168-5319
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | UMAB205 |
Immunogen | Full length human recombit protein of human TNNT2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cardiac muscle troponin T; cardiomyopathy, dilated 1D (autosomal dominant); cardiomyopathy, hypertrophic 2; CMD1D CMH2 CMPD2 LVNC6 RCM3 TnTC cTnT; cTnT; TnTc; troponin T type 2 (cardiac); Troponin T, cardiac muscle; troponin T2, cardiac; truncated cardiac troponin T
Gene Aliases: CMD1D; CMH2; CMPD2; cTnT; LVNC6; RCM3; TNNT2; TnTC
UniProt ID: (Human) P45379
Entrez Gene ID: (Human) 7139
Molecular Function:
actin binding motor protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.