Invitrogen
Manufacturer Code:PA587835
Catalog # PIPA587835
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-262 of human TNNT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: skeletal muscle slow-twitch TnT; Slow skeletal muscle troponin T; sTnT; TnTs; troponin T type 1 (skeletal, slow); Troponin T, slow skeletal muscle; troponin T1, skeletal, slow; troponin-T1, skeletal, slow
Gene Aliases: ANM; AW146156; Fang2; NEM5; ssTnT; STNT; TNNT1; TNT; TNTS
UniProt ID: (Human) P13805, (Rat) Q7TNB2, (Mouse) O88346
Entrez Gene ID: (Human) 7138, (Rat) 171409, (Mouse) 21955
Molecular Function:
actin binding motor protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.