Invitrogen
Manufacturer Code:PA521989
Catalog # PIPA521989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment contain a sequence corresponding to a region within amino acids 187 and 403 (Q59H18) of TNNI3K |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cardiac ankyrin repeat kinase; Cardiac troponin I-interacting kinase; CARK TNNI3-interacting kinase cardiac ankyrin repeat kinase cardiac troponin I-interacting kinase serine/threonine-protein kinase TNNI3K; Serine/threonine-protein kinase TNNI3K; TNNI3-interacting kinase; TNNI3K
Gene Aliases: CARK; CCDD; TNNI3K
UniProt ID: (Human) Q59H18
Entrez Gene ID: (Human) 51086
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.