Novus Biologicals
Manufacturer Code:NBP132837
Catalog # NBP132837
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human TNNI3K. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cardiac ankyrin repeat kinase; Cardiac troponin I-interacting kinase; CARKCardiac ankyrin repeat kinase EC 2.7.11.1 MGC142099 MGC33828 serine/threonine-protein kinase TNNI3K TNNI3 interacting kinase TNNI3-interacting kinase; Serine/threonine-protein kinase TNNI3K; TNNI3-interacting kinase; TNNI3K
Gene Aliases: CARK; CCDD; TNNI3K
UniProt ID: (Human) Q59H18
Entrez Gene ID: (Human) 51086
Molecular Function: kinase non-receptor serine/threonine protein kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.