Proteintech
Manufacturer Code:222531AP150UL
Catalog # 50-173-3798
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TNNI1 Fusion Protein Ag17749 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SSTNI TNN1 TNNI1; troponin I type 1 (skeletal, slow); troponin I, skeletal, slow 1; Troponin I, slow isoform; Troponin I, slow skeletal muscle; Troponin I, slow-twitch isoform
Gene Aliases: 2700018B22Rik; AI747285; SSTNI; TNN1; TNNI1
UniProt ID: (Human) P19237, (Mouse) Q9WUZ5, (Rat) P13413
Entrez Gene ID: (Human) 7135, (Mouse) 21952, (Rat) 29388
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.