OriGene
Manufacturer Code:UM800096
Catalog # 50-168-5316
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | UMAB204 |
Immunogen | Full length human recombit protein of human TNNC1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cardiac troponin C; CMD1Z CMH13 TN-C TNC TNNC; slow twitch skeletal/cardiac muscle troponin C; TN-C; troponin C type 1 (slow); Troponin C, slow skeletal and cardiac muscles; troponin C1, slow
Gene Aliases: CMD1Z; CMH13; TN-C; TNC; TNNC; TNNC1
UniProt ID: (Human) P63316
Entrez Gene ID: (Human) 7134
Molecular Function: actin family cytoskeletal protein calcium-binding protein calmodulin cytoskeletal protein intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.