Abnova
Manufacturer Code:PAB18670
Catalog # 89-120-227
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids at internal region of human TNKS2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of TNKS2.
For Research Use Only
Protein Aliases: ADP-ribosyltransferase diphtheria toxin-like 6; ARTD6; PARP-5b; PARP-5c; PARP5B; PARP5C; Poly [ADP-ribose] polymerase 5B; Poly [ADP-ribose] polymerase tankyrase-2; Protein poly-ADP-ribosyltransferase tankyrase-2; TANK2; Tankyrase II; tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase 2; Tankyrase-2; Tankyrase-like protein; Tankyrase-related protein; TNKL; TNKS-2; TRF1-interacting ankyrin-related ADP-ribose polymerase 2
Gene Aliases: ARTD6; PARP-5b; PARP-5c; PARP5B; PARP5C; pART6; TANK2; TNKL; TNKS2
UniProt ID: (Human) Q9H2K2
Entrez Gene ID: (Human) 80351
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.