Abnova
Manufacturer Code:PAB15673
Catalog # 89-117-738
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to N-terminus 120 amino acids of human TNKS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised aganist partial recombinant TNKS.
For Research Use Only
Protein Aliases: ADP-ribosyltransferase diphtheria toxin-like 5; ARTD5; PARP-5a; PARP5A; PARPL; Poly [ADP-ribose] polymerase 5A; Poly [ADP-ribose] polymerase tankyrase-1; Protein poly-ADP-ribosyltransferase tankyrase-1; TANK1; Tankyrase I; tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase; Tankyrase-1; TIN1; TINF1; TNKS-1; TNKS1; TRF1-interacting ankyrin-related ADP-ribose polymerase; TRF1-interacting ankyrin-related ADP-ribose polymerase 1
Gene Aliases: ARTD5; PARP-5a; PARP5A; PARPL; pART5; TIN1; TINF1; TNKS; TNKS1
UniProt ID: (Human) O95271
Entrez Gene ID: (Human) 8658
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.