Abnova
Manufacturer Code:H00008995M27
Catalog # 89-300-187
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3A4 |
Immunogen | TNFSF18 (AAH69111.1 50 a.a. ~ 177 a.a) partial recombinant protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Activation-inducible TNF-related ligand; AITR ligand; AITRL; GITR ligand; GITRL; Glucocorticoid-induced TNF-related ligand; glucocorticoid-induced TNFR-related protein ligand; hGITRL; MGC138237; TL6; tumor necrosis factor (ligand) superfamily member 18; tumor necrosis factor (ligand) superfamily, member 18; tumor necrosis factor ligand 2A; Tumor necrosis factor ligand superfamily member 18
Gene Aliases: AITRL; GITRL; hGITRL; TL6; TNFSF18; TNLG2A; UNQ149/PRO175
UniProt ID: (Human) Q9UNG2
Entrez Gene ID: (Human) 8995
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.