Abnova
Manufacturer Code:H00009966M09
Catalog # 89-300-016
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4F1 |
Immunogen | TNFSF15 (AAH74941.1 72 a.a. ~ 251 a.a) partial recombinant protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MGC129934; MGC129935; TL1; TL1A; TNF ligand-related molecule 1; TNF superfamily ligand TL1A; tumor necrosis factor (ligand) superfamily member 15; tumor necrosis factor (ligand) superfamily, member 15; tumor necrosis factor ligand 1B; Tumor necrosis factor ligand superfamily member 15; Tumor necrosis factor ligand superfamily member 15, membrane form; Tumor necrosis factor ligand superfamily member 15, secreted form; Vascular endothelial cell growth inhibitor; vascular endothelial growth inhibitor-192A; VEGI; VEGI192A
Gene Aliases: TL1; TL1A; TNFSF15; TNLG1B; VEGI; VEGI192A
UniProt ID: (Human) O95150
Entrez Gene ID: (Human) 9966
Molecular Function:
cytokine
signaling molecule
tumor necrosis factor family member
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.