Invitrogen
Manufacturer Code:MA515726
Catalog # PIMA515726
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5A11 |
Immunogen | Purified recombit fragment of human TNFRSF11B expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: OPG TR1 OCIF MGC29565 TNFRSF11B; Osteoclastogenesis inhibitory factor; Osteoprotegerin; Tumor necrosis factor receptor superfamily member 11B; tumor necrosis factor receptor superfamily, member 11b
Gene Aliases: OCIF; OPG; PDB5; TNFRSF11B; TR1
UniProt ID: (Human) O00300
Entrez Gene ID: (Human) 4982
Molecular Function:
cytokine receptor
receptor
tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.