Abnova
Manufacturer Code:MAB1297M03
Catalog # 89-300-312
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1H7 |
Immunogen | TNFRSF10A (AAH12866.1 24 a.a. ~ 239 a.a) partial recombinant protein with mouse IgG2a-Fc tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: APO2; CD261; cytotoxic TRAIL receptor; Death receptor 4; DR4; MGC9365; TNF-related apoptosis-inducing ligand receptor 1; TRAIL receptor 1; TRAIL-R1; TRAILR-1; TRAILR1; Tumor necrosis factor receptor superfamily member 10A; tumor necrosis factor receptor superfamily member 10a variant 2; tumor necrosis factor receptor superfamily, member 10a
Gene Aliases: APO2; CD261; DR4; TNFRSF10A; TRAILR-1; TRAILR1
UniProt ID: (Human) O00220
Entrez Gene ID: (Human) 8797
Molecular Function: cytokine receptor receptor tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.