Novus Biologicals
Manufacturer Code:NBP233522
Catalog # NBP233522
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: RINHVFNHFADVEFLSTLYSLDGDCRPNLKRICEGINKLLDEKVL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: TNF alpha-induced protein 8-like protein 3; TNF Alpha-Induced Protein 8-Like Protein 3 TNFAIP8-Like Protein 3 Tumor Necrosis Factor Alpha-Induced Protein 8-Like Protein 3 Tumor Necrosis Factor Alpha-Induced Protein 8-Like 3 Tumor Necrosis Factor Alpha-Induced Protein 8-Like Protein 3; TNFAIP8-like protein 3; Tumor necrosis factor alpha-induced protein 8-like protein 3; tumor necrosis factor, alpha induced protein 8 like 3; tumor necrosis factor, alpha-induced protein 8-like 3
Gene Aliases: TIPE3; TNFAIP8L3
UniProt ID: (Human) Q5GJ75
Entrez Gene ID: (Human) 388121
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.