Proteintech
Manufacturer Code:234561AP150UL
Catalog # 50-173-4081
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TNFAIP3 Fusion Protein Ag18150 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A20 OTUD7C TNF alpha induced protein 3 TNFA1P2 TNFAIP3 Zinc finger protein A20; A20p37; A20p50; OTU domain-containing protein 7C; Putative DNA-binding protein A20; TNF alpha-induced protein 3; Tumor necrosis factor alpha-induced protein 3; tumor necrosis factor induced protein 3; tumor necrosis factor inducible protein A20; tumor necrosis factor, alpha induced protein 3; tumor necrosis factor, alpha-induced protein 3; Zinc finger protein A20
Gene Aliases: A20; AISBL; OTUD7C; TNFA1P2; TNFAIP3; Tnfip3
UniProt ID: (Human) P21580, (Mouse) Q60769
Entrez Gene ID: (Human) 7128, (Rat) 683206, (Mouse) 21929
Molecular Function:
DNA binding protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.