Invitrogen
Manufacturer Code:PA565073
Catalog # PIPA565073
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human TNFAIP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: exocyst complex component 3-like 3; Primary response gene B94 protein; TNF alpha-induced protein 2; TNFAIP2 Tumor necrosis factor alpha-induced protein 2 exocyst complex component 3-like 3 EXOC3L3 B94; Tumor necrosis factor alpha-induced protein 2; tumor necrosis factor, alpha induced protein 2; tumor necrosis factor, alpha-induced protein 2
Gene Aliases: B94; EXOC3L3; TNFAIP2
UniProt ID: (Human) Q03169
Entrez Gene ID: (Human) 7127
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.