Invitrogen
Manufacturer Code:PA5104529
Catalog # PIPA5104529
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TNAP2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B94 Exoc3l3 exocyst complex component 3-like 3 M-sec Primary response gene B94 protein TNF alpha induced protein 2 TNF alpha-induced protein 2 TNF alpha-IP 2 Tnfaip2 tnfb94 Tnfip2 Tumor necrosis factor alpha-induced protein 2 tumor necrosis factor induced protein 2 tumor necrosis factor alpha induced protein 2 tumor necrosis factor alpha-induced protein 2; exocyst complex component 3-like 3; Primary response gene B94 protein; TNF alpha-induced protein 2; Tumor necrosis factor alpha-induced protein 2; tumor necrosis factor induced protein 2; tumor necrosis factor, alpha induced protein 2; tumor necrosis factor, alpha-induced protein 2
Gene Aliases: B94; EXOC3L3; M-sec; TNFAIP2; tnfb94; Tnfip2
UniProt ID: (Human) Q03169, (Mouse) Q61333
Entrez Gene ID: (Human) 7127, (Mouse) 21928
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.