Invitrogen
Manufacturer Code:PA5101162
Catalog # PIPA5101162
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TNAP1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B12 B61 BACURD2 BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 2 BTB/POZ domain-containing protein TNFAIP1 BTBD34 EDP1 Edp-1 hBACURD2 Protein B12 TNF alpha induced protein 1 TNFAIP1 Tnfip1 tumor necrosis factor induced protein 1 tumor necrosis factor alpha induced protein 1 tumor necrosis factor alpha-induced protein 1 (endothelial) Tumor necrosis factor alpha-induced protein 1 endothelial Tumor necrosis factor-induced protein 1; BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 2; BTB/POZ domain-containing protein TNFAIP1; hBACURD2; Protein B12; tumor necrosis factor induced protein 1; tumor necrosis factor, alpha induced protein 1; tumor necrosis factor, alpha-induced protein 1 (endothelial); Tumor necrosis factor, alpha-induced protein 1, endothelial; Tumor necrosis factor-induced protein 1
Gene Aliases: B12; B61; BACURD2; BTBD34; Edp-1; EDP1; hBACURD2; TNFAIP1; Tnfip1
UniProt ID: (Human) Q13829, (Rat) Q7TNY1, (Mouse) O70479
Entrez Gene ID: (Human) 7126, (Rat) 287543, (Mouse) 21927
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.