Invitrogen
Manufacturer Code:PA586571
Catalog # PIPA586571
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 31-80 of Human TNF-beta. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LT-alpha; lymphotoxin A; lymphotoxin alpha (TNF superfamily, member 1); Lymphotoxin-alpha; Lymphotoxin-alpha LT-alpha TNF-beta Lymphotoxin alpha LTalpha TNFbeta Tumor necrosis factor ligand superfamily member 1 LTA TNFB TNFSF1; TNF superfamily, member 1; TNF-beta; tumor necrosis factor beta; tumor necrosis factor ligand 1E; Tumor necrosis factor ligand superfamily member 1
Gene Aliases: LT; LTA; TNFB; TNFSF1; TNLG1E
UniProt ID: (Human) P01374, (Rat) Q06332
Entrez Gene ID: (Human) 4049, (Rat) 25008
Molecular Function:
cytokine
signaling molecule
tumor necrosis factor family member
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.