Invitrogen
Manufacturer Code:PA5116055
Catalog # PIPA5116055
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TNF beta corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAMA-25N12.13 LT LTA LT-alpha lymphotoxin A lymphotoxin alpha lymphotoxin alpha (TNF superfamily member 1) lymphotoxin-alpha TNF b TNF beta TNF superfamily member 1 TNF TNFB TNFbeta TNF-beta TNFSF1 TNF TNLG1E tumor necrosis factor beta tumor necrosis factor ligand 1E tumor necrosis factor ligand superfamily member 1; LT-alpha; lymphotoxin A; lymphotoxin alpha; lymphotoxin alpha (TNF superfamily, member 1); Lymphotoxin-alpha; TNF beta; TNF superfamily, member 1; TNF-beta; tumor necrosis factor beta; tumor necrosis factor ligand 1E; Tumor necrosis factor ligand superfamily member 1
Gene Aliases: hlb382; LT; LT-[a]; LT-alpha; LT[a]; LTA; LTalpha; Ltx; TNF-beta; TNFB; TNFSF1; Tnfsf1b; TNLG1E
UniProt ID: (Human) P01374, (Mouse) P09225, (Rat) Q06332
Entrez Gene ID: (Human) 4049, (Mouse) 16992, (Rat) 25008
Molecular Function:
cytokine
signaling molecule
tumor necrosis factor family member
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.