Proteintech
Manufacturer Code:602911IG150UL
Catalog # 50-173-6386
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7B8A11 |
Immunogen | TNF alpha Fusion Protein Ag11413 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: APC1 protein; C-domain 1; C-domain 2; Cachectin; Cachectin DIF TNF TNF a TNF alpha TNFA TNF-a TNFSF2 TNFalpha Tumor necrosis factor; ICD1; ICD2; Intracellular domain 1; Intracellular domain 2; N-terminal fragment; NTF; TNF, macrophage-derived; TNF, monocyte-derived; TNF-a; TNF-alpha; Tumor necrosis factor; tumor necrosis factor ligand 1F; Tumor necrosis factor ligand superfamily member 2; Tumor necrosis factor, membrane form; Tumor necrosis factor, soluble form; tumor necrosis factor-alpha
Gene Aliases: DIF; TNF; TNF-a; TNF-alpha; TNFA; TNFalpha; Tnfsf1a; TNFSF2; TNLG1F
UniProt ID: (Human) P01375, (Mouse) P06804
Entrez Gene ID: (Human) 7124, (Mouse) 21926
Molecular Function: cytokine signaling molecule tumor necrosis factor family member
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.