Novus Biologicals
Manufacturer Code:NBP275925
Catalog # NBP275925
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2342C |
Immunogen | Partial recombinant human TNF-alpha protein (amino acids 77-233) [Uniprot: P01375] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: APC1 protein; APC1 protein Cachectin DIF TNF monocyte-derived TNF-a TNF-alpha TNF-alpha/TNFSF2 TNF-alphacachectin TNFATNF macrophage-derived TNFSF2TNF superfamily member 2 tumor necrosis factor tumor necrosis factor (TNF superfamily member 2) tumor necrosis factor alpha Tumor necrosis factor ligand superfamily member 2 tumor necrosis factor-alpha; C-domain 1; C-domain 2; Cachectin; ICD1; ICD2; Intracellular domain 1; Intracellular domain 2; N-terminal fragment; NTF; TNF, macrophage-derived; TNF, monocyte-derived; TNF-a; TNF-alpha; Tumor necrosis factor; tumor necrosis factor ligand 1F; Tumor necrosis factor ligand superfamily member 2; Tumor necrosis factor, membrane form; Tumor necrosis factor, soluble form; tumor necrosis factor-alpha
Gene Aliases: DIF; TNF; TNF-alpha; TNFA; TNFSF2; TNLG1F
UniProt ID: (Human) P01375
Entrez Gene ID: (Human) 7124
Molecular Function:
cytokine
signaling molecule
tumor necrosis factor family member
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.