Proteintech
Manufacturer Code:210401AP150UL
Catalog # 50-173-3221
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TMX3 Fusion Protein Ag15376 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1830 PDIA13 TMX3 TXNDC10; protein disulfide isomerase family A, member 13; Protein disulfide-isomerase TMX3; thioredoxin domain containing 10; Thioredoxin domain-containing protein 10; Thioredoxin-related transmembrane protein 3
Gene Aliases: 6430411B10Rik; A730024F05Rik; AV259382; KIAA1830; mKIAA1830; PDIA13; TMX3; TXNDC10
UniProt ID: (Human) Q96JJ7, (Mouse) Q8BXZ1
Entrez Gene ID: (Human) 54495, (Mouse) 67988
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.