Invitrogen
Manufacturer Code:PA543989
Catalog # PIPA543989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human TMPRSS6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: matriptase 2; Matriptase-2; membrane-bound mosaic serine proteinase matriptase-2; Transmembrane protease serine 6; Transmembrane protease serine 6 IRIDA; transmembrane protease, serine 6; transmembrane serine protease 6; type II transmembrane serine protease 6
Gene Aliases: 1300008A22Rik; IRIDA; TMPRSS6; UNQ354/PRO618
UniProt ID: (Human) Q8IU80, (Mouse) Q9DBI0
Entrez Gene ID: (Dog) 608593, (Pig) 100516673, (Human) 164656, (Horse) 100069647, (Rat) 315388, (Mouse) 71753, (Guinea pig) 100730361
Molecular Function: annexin calcium-binding protein calmodulin enzyme modulator hydrolase intracellular calcium-sensing protein peptide hormone protease protease inhibitor receptor serine protease signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.