Proteintech
Manufacturer Code:129501AP150UL
Catalog # 50-172-8371
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TMPRSS6 Fusion Protein Ag4076 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IRIDA Matriptase 2 TMPRSS6; matriptase 2; Matriptase-2; membrane-bound mosaic serine proteinase matriptase-2; Transmembrane protease serine 6; transmembrane protease, serine 6; transmembrane serine protease 6; type II transmembrane serine protease 6
Gene Aliases: 1300008A22Rik; IRIDA; TMPRSS6; UNQ354/PRO618
UniProt ID: (Human) Q8IU80, (Mouse) Q9DBI0
Entrez Gene ID: (Human) 164656, (Rat) 315388, (Mouse) 71753
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.