Abnova
Manufacturer Code:H00164656K
Catalog # 89-208-543
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human TMPRSS6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human TMPRSS6 peptide using ARM Technology.
For Research Use Only
Protein Aliases: IRIDA; Matriptase-2; membrane-bound mosaic serine proteinase matriptase-2; Transmembrane protease serine 6; type II transmembrane serine protease 6
Gene Aliases: IRIDA; TMPRSS6; UNQ354/PRO618
UniProt ID: (Human) Q8IU80
Entrez Gene ID: (Human) 164656
Molecular Function: annexin calcium-binding protein calmodulin enzyme modulator hydrolase intracellular calcium-sensing protein peptide hormone protease protease inhibitor receptor serine protease signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.