Proteintech
Manufacturer Code:179531AP150UL
Catalog # 50-173-1897
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TMPRSS3 Fusion Protein Ag12435 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DFNB10 DFNB8 ECHOS1 Serine protease TADG 12 TADG12 TMPRSS3; Serine protease TADG-12; Transmembrane protease serine 3; transmembrane proteinase serine 3; Tumor-associated differentially-expressed gene 12 protein
Gene Aliases: DFNB10; DFNB8; ECHOS1; TADG12; TMPRSS3; UNQ323/PRO382
UniProt ID: (Human) P57727, (Mouse) Q8K1T0
Entrez Gene ID: (Human) 64699, (Mouse) 140765, (Rat) 309665
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.