Invitrogen
Manufacturer Code:PA520316
Catalog # PIPA520316
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | An 18 amino acid peptide from near the center of human TMP21. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 21 kDa transmembrane trafficking protein; 21 kDa transmembrane-trafficking protein; integral membrane protein Tmp21-I (p23); p23; p24 family protein delta-1; P24(DELTA) S31I125 S31III125 TMP21 Tmp-21-I p23 21 kDa transmembrane trafficking protein 21 kDa transmembrane-trafficking protein p24 family protein delta-1 p24delta p24delta1 transmembrane emp24 domain-containing protein 10 transmembrane protein Tmp21; p24delta; p24delta1; Protein TMED10; S31I125; S31III125; testicular tissue protein Li 206; Tmp-21-I; Transmembrane emp24 domain-containing protein 10; transmembrane emp24-like trafficking protein 10; Transmembrane protein Tmp21; transmembrane trafficking protein; transmembrane trafficking protein 21
Gene Aliases: 1110014C03Rik; p23; P24(DELTA); p24d1; p24delta1; S31I125; S31III125; TMED10; Tmp-21-I; TMP21
UniProt ID: (Human) P49755, (Rat) Q63584, (Mouse) Q9D1D4
Entrez Gene ID: (Human) 10972, (Rat) 84599, (Mouse) 68581
Molecular Function:
membrane traffic protein
transfer/carrier protein
vesicle coat protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.