Invitrogen
Manufacturer Code:PA146005
Catalog # 01-674-583
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic internal peptide corresponding to residues 50-150 of the human TMP21 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 21 kDa transmembrane trafficking protein; 21 kDa transmembrane-trafficking protein; Integral membrane protein p23; integral membrane protein Tmp21-I (p23); p23; p24 family protein delta-1; p24delta; p24delta1; Protein TMED10; S31I125; S31III125; testicular tissue protein Li 206; Tmp-21-I; Transmembrane emp24 domain-containing protein 10; Transmembrane emp24 domain-containing protein 10 Transmembrane protein Tmp21 21 kDa transmembrane trafficking prote; transmembrane emp24-like trafficking protein 10; Transmembrane protein Tmp21; transmembrane trafficking protein; transmembrane trafficking protein 21
Gene Aliases: 1110014C03Rik; p23; P24(DELTA); p24d1; p24delta1; S31I125; S31III125; TMED10; Tmp-21-I; TMP21; XELAEV_18000191mg; Xp24d2; Xp24delta2
UniProt ID: (Bovine) Q5E971, (Human) P49755, (Rabbit) Q28735, (Rat) Q63584, (Mouse) Q9D1D4
Entrez Gene ID: (Bovine) 529761, (Human) 10972, (Rabbit) 100009296, (Rat) 84599, (Mouse) 68581, (Xenopus) 379442
Molecular Function: membrane traffic protein transfer/carrier protein vesicle coat protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.