Invitrogen
Manufacturer Code:PA551645
Catalog # PIPA551645
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human TMOD3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: tropomodulin 3 (ubiquitous); tropomodulin 3; ubiquitous tropomodulin; Tropomodulin-3; U-Tmod; Ubiquitous tropomodulin; UTMOD tropomodulin-3 ubiquitous tropomodulin
Gene Aliases: TMOD3; U-Tmod; UTMOD
UniProt ID: (Human) Q9NYL9, (Mouse) Q9JHJ0
Entrez Gene ID: (Human) 29766, (Mouse) 50875, (Rat) 300838
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.