Proteintech
Manufacturer Code:125211AP150UL
Catalog # 50-172-8038
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TMOD3 Fusion Protein Ag3215 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: TMOD3 Tropomodulin 3 tropomodulin 3 (ubiquitous) U Tmod Ubiquitous tropomodulin UTMOD; tropomodulin 3 (ubiquitous); tropomodulin 3; ubiquitous tropomodulin; Tropomodulin-3; U-Tmod; Ubiquitous tropomodulin
Gene Aliases: TMOD3; U-Tmod; UTMOD
UniProt ID: (Human) Q9NYL9, (Mouse) Q9JHJ0
Entrez Gene ID: (Human) 29766, (Mouse) 50875, (Rat) 300838
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.