Invitrogen
Manufacturer Code:PA552598
Catalog # PIPA552598
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human TMF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Androgen receptor coactivator 160 kDa protein; Androgen receptor-associated protein of 160 kDa; ARA160 TMF TATA element modulatory factor androgen receptor coactivator 160 kDa protein androgen receptor-associated protein of 160 kDa; TATA element modulatory factor; TMF
Gene Aliases: 7030402D04Rik; ARA160; Gm153; TMF; TMF1; Tmf1`
UniProt ID: (Human) P82094, (Mouse) B9EKI3
Entrez Gene ID: (Human) 7110, (Rat) 114206, (Mouse) 232286
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.