Invitrogen
Manufacturer Code:PA542676
Catalog # PIPA542676
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human TMEM91 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dispanin subfamily C member 3; DSPC3; interferon induced transmembrane protein domain containing 6; Transmembrane protein 91
Gene Aliases: DSPC3; IFITMD6; TMEM91
UniProt ID: (Human) Q6ZNR0
Entrez Gene ID: (Human) 641649, (Pig) 100511932, (Dog) 100682781, (Rat) 100359879
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.