Invitrogen
Manufacturer Code:PA569917
Catalog # PIPA569917
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of mouse TMEM86A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Lysoplasmalogenase-like protein TMEM86A; Transmembrane protein 86A
Gene Aliases: 1810054O13Rik; AI414959; AW742698; im:7137034; RGD1305749; TMEM86A; zgc:113571
UniProt ID: (Bovine) Q3MHQ7, (Human) Q8N2M4, (Mouse) Q9D8N3
Entrez Gene ID: (Bovine) 532018, (Human) 144110, (Horse) 100057256, (Rabbit) 100343684, (Mouse) 67893, (Guinea pig) 100735970, (Rat) 308602, (Zebrafish) 541385
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.