Invitrogen
Manufacturer Code:PA543489
Catalog # PIPA543489
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human TMEM79 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mattrin; Transmembrane protein 79
Gene Aliases: 2310042N02Rik; 2310074C17Rik; MATT; RGD1309886; TMEM79
UniProt ID: (Bovine) Q5E9U3, (Human) Q9BSE2, (Mouse) Q9D709, (Rat) Q3T1H8
Entrez Gene ID: (Rabbit) 100338752, (Bovine) 513599, (Dog) 612144, (Pig) 100152302, (Human) 84283, (Horse) 100057797, (Mouse) 71913, (Guinea pig) 100722654, (Rat) 310626
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.