R&D Systems
Manufacturer Code:MAB9118
Catalog # MAB9118
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 816004 |
Immunogen | <em>E.coli</em>-derived recombinant human TMEM59 Met1-Asn220 Accession # Q9BXS4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C1orf8 chromosome 1 open reading frame 8 DCF1 FLJ42977 HSPC001 Liver membrane-bound protein PRO195 transmembrane protein 59 UNQ169; dendritic cell factor 1; Liver membrane-bound protein; Transmembrane protein 59
Gene Aliases: C1orf8; DCF1; HSPC001; PRO195; TMEM59; UNQ169; UNQ169/PRO195
UniProt ID: (Human) Q9BXS4
Entrez Gene ID: (Human) 9528
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.