Invitrogen
Manufacturer Code:PA521575
Catalog # PIPA521575
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 16 and 311 of TMEM59 (Uniprot IDNo.Q9BXS4) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dendritic cell factor 1; Liver membrane-bound protein; RP4-758J24.2 C1orf8 HSPC001 liver membrane-bound protein; Thymic dendritic cell-derived factor 1; Transmembrane protein 59
Gene Aliases: 1110001M20Rik; 3110046P06Rik; AI256529; C1orf8; D4Ertd20e; DCF1; HSPC001; MTDCF1; ORF18; PRO195; RGD1310313; Tdcf1; TMEM59; UNQ169; UNQ169/PRO195
UniProt ID: (Human) Q9BXS4, (Mouse) Q9QY73
Entrez Gene ID: (Human) 9528, (Mouse) 56374, (Rat) 100196907
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.