Proteintech
Manufacturer Code:170951AP150UL
Catalog # 50-173-1381
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TMEM5 Fusion Protein Ag10562 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HP10481 TMEM5 transmembrane protein 5; Ribitol-5-phosphate xylosyltransferase 1; Transmembrane protein 5; type II membrane protein; UDP-D-xylose:ribitol-5-phosphate beta1,4-xylosyltransferase
Gene Aliases: 6330415D21Rik; HP10481; MDDGA10; RXYLT1; TMEM5
UniProt ID: (Human) Q9Y2B1, (Mouse) Q8VDX6
Entrez Gene ID: (Human) 10329, (Mouse) 216395, (Rat) 299841
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.