Invitrogen
Manufacturer Code:PA542844
Catalog # PIPA542844
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human TMEM48 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hNDC1; nuclear division cycle 1 homolog; Nucleoporin NDC1; Transmembrane protein 48; Transmembrane protein 48 FLJ10407 FLJ12556 FLJ34120 NDC1 RP4-654H19.1 NET3 TMEM48
Gene Aliases: 2810475A17Rik; AI450313; NDC1; NET3; TMEM48; wu:fk93f04; zgc:55636
UniProt ID: (Human) Q9BTX1, (Rat) Q6AXN4, (Mouse) Q8VCB1, (Zebrafish) Q7SZC5
Entrez Gene ID: (Human) 55706, (Horse) 100050513, (Rabbit) 100354365, (Dog) 100684187, (Pig) 100621612, (Rat) 362557, (Guinea pig) 100716231, (Mouse) 72787, (Zebrafish) 335236
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.