Invitrogen
Manufacturer Code:PA520859
Catalog # PIPA520859
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 17 amino acid peptide from near the carboxy terminus of human TMEM38B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mitsugumin-33B; Mitsugumin33B; RP11-219P18.1 C9orf87 D4Ertd89e TRIC-B TRICB bA219P18.1 trimeric intracellular cation channel type B; Transmembrane protein 38B; TRIC-B; TRICB; Trimeric intracellular cation channel type B
Gene Aliases: 1600017F22Rik; AA437809; AV051057; bA219P18.1; C9orf87; D4Ertd89e; Mg33b; OI14; RGD1305703; TMEM38B; TRIC-B; TRICB
UniProt ID: (Human) Q9NVV0, (Rat) Q68FV1, (Mouse) Q9DAV9
Entrez Gene ID: (Human) 55151, (Rat) 362521, (Mouse) 52076
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.