Invitrogen
Manufacturer Code:PA577754
Catalog # PIPA577754
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)DNHGAPHGMGLGTQHS corresponding to amino acid residues 259-274 of rat |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 27 kDa sarcoplasmic reticulum protein; Mitsugumin-33A; Mitsugumin33A; sarcoplasmic reticulum protein 27; SPR-27; tmem38a {ECO:0000250|UniProtKB:Q3TMP8}; Transmembrane protein 38A; TRIC-A; TRICA; Trimeric intracellular cation channel type A
Gene Aliases: 1110001E17Rik; AI413399; mg33a; RGD1307901; Srp-27; Tmem38a; TRIC-A
UniProt ID: (Mouse) Q3TMP8, (Rat) A6ZIQ8
Entrez Gene ID: (Mouse) 74166, (Rat) 306327
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.