Invitrogen
Manufacturer Code:PA5113797
Catalog # PIPA5113797
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human TMEM30B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9130011B11Rik CDC50B Cell cycle control protein 50B P4-ATPase flippase complex beta subunit TMEM30B Tmem30b Transmembrane protein 30B; Cell cycle control protein 50B; P4-ATPase flippase complex beta subunit TMEM30B; Transmembrane protein 30B
Gene Aliases: CDC50B; TMEM30B
UniProt ID: (Human) Q3MIR4
Entrez Gene ID: (Human) 161291
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.