Invitrogen
Manufacturer Code:PA543041
Catalog # PIPA543041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human TMEM30A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell cycle control protein 50A; Cell cycle control protein 50A-like protein; P4-ATPase flippase complex beta subunit TMEM30A; Transmembrane protein 30A
Gene Aliases: 2010200I23Rik; AW540225; C6orf67; CDC50A; D9Wsu20e; TMEM30A
UniProt ID: (Human) Q9NV96, (Bovine) Q17QL5, (Mouse) Q8VEK0, (Rat) Q6AY41
Entrez Gene ID: (Horse) 100068662, (Human) 55754, (Dog) 474973, (Bovine) 505797, (Mouse) 69981, (Guinea pig) 100715364, (Rat) 300857
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.