Invitrogen
Manufacturer Code:PA588721
Catalog # PIPA588721
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-222 of human TMEM27 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Collectrin; kidney-specific membrane protein; Transmembrane protein 27
Gene Aliases: 0610008J07Rik; Cltrn; collectrin; NX-17; NX17; Tmem27
UniProt ID: (Rat) Q9ESG3, (Mouse) Q9ESG4
Entrez Gene ID: (Rat) 57395, (Mouse) 57394
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.