Invitrogen
Manufacturer Code:PA547676
Catalog # PIPA547676
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Collectrin Glu15-Ile144 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Collectrin; kidney-specific membrane protein; NX17 TMEM27 collectrin kidney-specific membrane protein NX-17 transmembrane protein 27NX170610008J07Rik; Transmembrane protein 27
Gene Aliases: 0610008J07Rik; Cltrn; NX-17; NX17; Tmem27
UniProt ID: (Mouse) Q9ESG4
Entrez Gene ID: (Mouse) 57394
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.