Invitrogen
Manufacturer Code:PA569313
Catalog # PIPA569313
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human TMEM195 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGMO TMEM195; Alkylglycerol monooxygenase; glyceryl-ether monooxygenase; Transmembrane protein 195
Gene Aliases: A530016O06Rik; AGMO; AI790538; RGD1312038; TMEM195
UniProt ID: (Human) Q6ZNB7, (Rat) A0JPQ8, (Mouse) Q8BS35
Entrez Gene ID: (Horse) 100052771, (Human) 392636, (Dog) 475247, (Pig) 100513371, (Rabbit) 100339522, (Bovine) 613658, (Rat) 362732, (Mouse) 319660
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.