Proteintech
Manufacturer Code:554111AP150UL
Catalog # 50-173-6146
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 178; GPR178 KIAA1423 TMEM181 transmembrane protein 181; Transmembrane protein 181
Gene Aliases: 5930418K15Rik; C76977; GPR178; KIAA1423; mKIAA1423; TMEM181
UniProt ID: (Human) Q9P2C4
Entrez Gene ID: (Human) 57583, (Mouse) 77106
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.