Novus Biologicals
Manufacturer Code:NBP174231
Catalog # NBP174231
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to the N terminal of Tmem102. Immunizing peptide sequence KDFVFALLGLVHRQDPRFPPQAELLLLRGGIREGSLDLGHAPLGPYSRGP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CBAP; CBAP common beta-chain associated protein FLJ36878 transmembrane protein 102; Common beta-chain associated protein; common beta-chain-associated protein; Transmembrane protein 102
Gene Aliases: CBAP; TMEM102
UniProt ID: (Human) Q8N9M5
Entrez Gene ID: (Human) 284114
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.