Proteintech
Manufacturer Code:255051AP150UL
Catalog # 50-173-4937
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TMEM102 Fusion Protein Ag22213 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CBAP; CBAP TMEM102 transmembrane protein 102; Common beta-chain associated protein; common beta-chain-associated protein; Transmembrane protein 102; transmembrane protein 102, pseudogene
Gene Aliases: 4632403M07Rik; CBAP; TMEM102; Tmem102-ps
UniProt ID: (Human) Q8N9M5, (Mouse) Q3UPR7
Entrez Gene ID: (Human) 284114, (Mouse) 380705
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.